siRNA Details
| Gene Symbol: Gsg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759923.1 | XP_008758145.1 | 4 | 677 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsg1l| Gene Symbol: Gsg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759923.1 | XP_008758145.1 | 4 | 677 | ||