siRNA Details
| Gene Symbol: Nt5dc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134887.1 | NP_001128359.1 | 2 | 375 | ||
| XM_006241203.2 | XP_006241265.1 | 2 | 376 | |||
| XM_006241204.2 | XP_006241266.1 | 2 | 377 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nt5dc3| Gene Symbol: Nt5dc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134887.1 | NP_001128359.1 | 2 | 375 | ||
| XM_006241203.2 | XP_006241265.1 | 2 | 376 | |||
| XM_006241204.2 | XP_006241266.1 | 2 | 377 | |||