siRNA Details
| Gene Symbol: Mink1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271136.1 | NP_001258065.1 | 16 | 2205 | ||
| XM_006246716.2 | XP_006246778.1 | 17 | 2332 | |||
| XM_006246717.2 | XP_006246779.1 | 17 | 2332 | |||
| XM_006246718.2 | XP_006246780.1 | 17 | 2332 | |||
| XM_006246720.2 | XP_006246782.1 | 16 | 2245 | |||
| XM_006246721.2 | XP_006246783.1 | 17 | 2332 | |||
| XM_006246722.2 | XP_006246784.1 | 16 | 2245 | |||
| XM_006246723.2 | XP_006246785.1 | 16 | 2245 | |||
| XM_006246724.2 | XP_006246786.1 | 16 | 2245 | |||
| XM_006246725.2 | XP_006246787.1 | 16 | 2057 | |||

Mink1
Genomic Map