siRNA Details
| Gene Symbol: Pcgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129882.1 | NP_001123354.1 | 7 | 819 | ||
| XM_006231311.2 | XP_006231373.1 | 7 | 912 | |||
| XM_008760377.1 | XP_008758599.1 | 6 | 537 | |||
| GenBank mRNA | BC167067.1 | - | 7 | 819 | 7104558 | |
| FQ220230.1 | - | 5 | 410 | |||
| FQ233837.1 | - | 7 | 754 | |||

Pcgf5
Genomic Map