siRNA Details
| Gene Symbol: Traf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233999.2 | XP_006234061.1 | 3 | 307 | ||
| XM_006234000.2 | XP_006234062.1 | 3 | 222 | |||
| GenBank mRNA | BC158696.1 | - | 3 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traf1| Gene Symbol: Traf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006233999.2 | XP_006234061.1 | 3 | 307 | ||
| XM_006234000.2 | XP_006234062.1 | 3 | 222 | |||
| GenBank mRNA | BC158696.1 | - | 3 | 94 | ||