siRNA Details
| Gene Symbol: Vom2r38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099476.1 | NP_001092946.1 | 1399 | |||
| Gene Symbol: Vom2r37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099487.1 | NP_001092957.1 | 3 | 1180 | ||
More Information
Set Membership: |
|

Vom2r38
Genomic Map