siRNA Details
| Gene Symbol: FARP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282983.1 | NP_001269912.1 | 8 | 817 | ||
| NM_001282984.1 | NP_001269913.1 | 8 | 817 | |||
| NM_014808.3 | NP_055623.1 | 8 | 817 | |||
| XM_005247050.3 | XP_005247107.1 | 8 | 781 | |||
| XM_011512233.1 | XP_011510535.1 | 8 | 781 | |||
| XM_011512234.1 | XP_011510536.1 | 8 | 706 | |||
| XM_011512235.1 | XP_011510537.1 | 9 | 1039 | |||
| XM_011512236.1 | XP_011510538.1 | 8 | 835 | |||
| XM_017005416.1 | XP_016860905.1 | 9 | 996 | |||
| XM_017005417.1 | XP_016860906.1 | 9 | 996 | |||
| XM_017005418.1 | XP_016860907.1 | 6 | 634 | |||
| XM_017005419.1 | XP_016860908.1 | 8 | 781 | |||
| XM_017005420.1 | XP_016860909.1 | 8 | 781 | |||
| GenBank mRNA | AB018336.1 | - | 8 | 770 | ||
| AK300435.1 | - | 8 | 777 | |||
| BC021301.2 | - | 8 | 769 | 5013180 | ||

FARP2
Genomic Map