siRNA Details
Gene Symbol: Gsta4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106840.1 | NP_001100310.1 | 831 | |||
GenBank mRNA | BC157819.1 | - | 801 | 5621777 | ||
FQ230022.1 | - | 811 | ||||
FQ233840.1 | - | 813 | ||||
X62660.1 | - | 792 |
Gene Symbol: LOC100909835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750542.3 | XP_003750590.1 | 1 | 850 |
