siRNA Details
| Gene Symbol: LOC690483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228950.2 | XP_006229012.1 | 3 | 941 | ||
| GenBank mRNA | BC082038.1 | - | 3 | 765 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC690483| Gene Symbol: LOC690483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228950.2 | XP_006229012.1 | 3 | 941 | ||
| GenBank mRNA | BC082038.1 | - | 3 | 765 | ||