siRNA Details
| Gene Symbol: Wscd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271178.1 | NP_001258107.1 | 2 | 595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wscd2| Gene Symbol: Wscd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271178.1 | NP_001258107.1 | 2 | 595 | ||