siRNA Details
| Gene Symbol: Tmco5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234788.2 | XP_006234850.1 | 12 | 902 | ||
| XM_006234789.2 | XP_006234851.1 | 12 | 882 | |||
| XM_006234790.2 | XP_006234852.1 | 12 | 927 | |||
| XM_006234791.2 | XP_006234853.1 | 11 | 784 | |||
| XM_008762112.1 | XP_008760334.1 | 12 | 959 | |||

Tmco5a
Genomic Map