siRNA Details
| Gene Symbol: LOC501233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047967.1 | NP_001041432.1 | 3 | 221 | ||
| GenBank mRNA | AY387066.1 | - | 3 | 221 | ||
| Gene Symbol: Khdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766940.1 | XP_008765162.1 | 1 | 377 | ||

LOC501233
Genomic Map