siRNA Details
| Gene Symbol: LOC100910827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CB775857.1 | - | 2 | 131 | ||
| FM054489.1 | - | 2 | 119 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100910827| Gene Symbol: LOC100910827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CB775857.1 | - | 2 | 131 | ||
| FM054489.1 | - | 2 | 119 | |||