siRNA Details
| Gene Symbol: Pla2g4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234810.2 | XP_006234872.1 | 8 | 726 | ||
| XM_008762122.1 | XP_008760344.1 | 8 | 726 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g4f| Gene Symbol: Pla2g4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234810.2 | XP_006234872.1 | 8 | 726 | ||
| XM_008762122.1 | XP_008760344.1 | 8 | 726 | |||