siRNA Details
| Gene Symbol: Wdr76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234874.2 | XP_006234936.1 | 3 | 771 | ||
| XM_006234875.2 | XP_006234937.1 | 3 | 771 | |||
| XM_006234876.2 | XP_006234938.1 | 2 | 216 | |||
| XM_006234877.1 | XP_006234939.1 | 2 | 209 | |||
| XM_230512.7 | XP_230512.4 | 2 | 247 | |||

Wdr76
Genomic Map