siRNA Details
| Gene Symbol: Tmem239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749572.3 | XP_003749620.1 | 1 | 189 | ||
| XM_006235049.2 | XP_006235111.1 | 1 | 241 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem239| Gene Symbol: Tmem239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749572.3 | XP_003749620.1 | 1 | 189 | ||
| XM_006235049.2 | XP_006235111.1 | 1 | 241 | |||