siRNA Details
| Gene Symbol: Tcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271277.1 | NP_001258206.1 | 2 | 863 | ||
| GenBank mRNA | FQ215695.1 | - | 2 | 861 | ||
| FQ216780.1 | - | 2 | 863 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcap| Gene Symbol: Tcap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271277.1 | NP_001258206.1 | 2 | 863 | ||
| GenBank mRNA | FQ215695.1 | - | 2 | 861 | ||
| FQ216780.1 | - | 2 | 863 | |||