siRNA Details
| Gene Symbol: Krtap3-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768204.1 | XP_008766426.1 | 1 | 279 | ||
| Gene Symbol: Krtap3-3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247444.2 | XP_006247506.1 | 1 | 286 | ||

Krtap3-3
Genomic Map