siRNA Details
| Gene Symbol: Cfhr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001068704.5 | XP_001068704.3 | 2 | 285 | ||
| XM_006249947.2 | XP_006250009.1 | 2 | 286 | |||
| XM_006249948.2 | XP_006250010.1 | 2 | 286 | |||
| XM_006249949.2 | XP_006250011.1 | 2 | 285 | |||
| XM_006249950.2 | XP_006250012.1 | 2 | 285 | |||
| XM_006249951.2 | XP_006250013.1 | 2 | 286 | |||

Cfhr2
Genomic Map