siRNA Details
| Gene Symbol: Tcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035237.1 | NP_001030314.1 | 14 | 1334 | ||
| NM_133524.2 | NP_598208.2 | 14 | 1334 | |||
| GenBank mRNA | AJ973227.1 | - | 14 | 1334 | ||
| BC078750.1 | - | 14 | 1262 | 7123064 | ||
| X54549.1 | - | 14 | 1334 | |||
| X62323.1 | - | 14 | 1238 | |||

Tcf3
Genomic Map