siRNA Details
| Gene Symbol: Xlr3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229630.2 | XP_006229692.1 | 9 | 1735 | ||
| XM_008759622.1 | XP_008757844.1 | 9 | 1828 | |||
| Gene Symbol: LOC100911047 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229628.2 | XP_006229690.1 | 9 | 1736 | ||
| XM_008759621.1 | XP_008757843.1 | 9 | 1829 | |||

Xlr3a
Genomic Map