siRNA Details
| Gene Symbol: Kif26a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240635.2 | XP_006240697.1 | 16 | 6100 | ||
| XM_006240636.2 | XP_006240698.1 | 16 | 6097 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif26a| Gene Symbol: Kif26a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240635.2 | XP_006240697.1 | 16 | 6100 | ||
| XM_006240636.2 | XP_006240698.1 | 16 | 6097 | |||