siRNA Details
| Gene Symbol: Tmc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126301.1 | NP_001119773.1 | 5 | 701 | ||
| XM_006247818.2 | XP_006247880.1 | 5 | 1149 | |||
| XM_006247819.2 | XP_006247881.1 | 5 | 1146 | |||
| XM_006247821.2 | XP_006247883.1 | 5 | 683 | |||
| XM_008768392.1 | XP_008766614.1 | 5 | 674 | |||
| GenBank mRNA | BC160873.1 | - | 5 | 701 | 7366375 | |

Tmc8
Genomic Map