siRNA Details
| Gene Symbol: LOC688286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247853.2 | XP_006247915.1 | 3 | 1985 | ||
| XM_006247854.2 | XP_006247916.1 | 5 | 1239 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC688286| Gene Symbol: LOC688286 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247853.2 | XP_006247915.1 | 3 | 1985 | ||
| XM_006247854.2 | XP_006247916.1 | 5 | 1239 | |||