siRNA Details
| Gene Symbol: Efemp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277341.1 | NP_001264270.1 | 5 | 577 | ||
| XM_006230742.2 | XP_006230804.1 | 5 | 917 | |||
| XM_006230743.1 | XP_006230805.1 | 5 | 568 | |||
| XM_006230744.2 | XP_006230806.1 | 5 | 574 | |||

Efemp2
Genomic Map