siRNA Details
| Gene Symbol: Aatk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168703.1 | NP_001162174.1 | 2, 3 | 331 | ||
| XM_006247926.2 | XP_006247988.1 | 2, 3 | 443 | |||
| XM_006247928.2 | XP_006247990.1 | 2, 3 | 335 | |||
| XM_008768484.1 | XP_008766706.1 | 2, 3 | 356 | |||

Aatk
Genomic Map