siRNA Details
| Gene Symbol: NEWGENE_1590060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768495.1 | XP_008766717.1 | 2 | 339 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEWGENE_1590060| Gene Symbol: NEWGENE_1590060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768495.1 | XP_008766717.1 | 2 | 339 | ||