siRNA Details
| Gene Symbol: Stra13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750986.3 | XP_003751034.2 | 2 | 224 | ||
| XM_008768490.1 | XP_008766712.1 | 2 | 236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stra13| Gene Symbol: Stra13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750986.3 | XP_003751034.2 | 2 | 224 | ||
| XM_008768490.1 | XP_008766712.1 | 2 | 236 | |||