siRNA Details
Gene Symbol: Rac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768496.1 | XP_008766718.1 | 5, 6 | 1036 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768496.1 | XP_008766718.1 | 5, 6 | 1036 |