siRNA Details
| Gene Symbol: LOC689629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760982.1 | XP_008759204.1 | 2 | 580 | ||
| Gene Symbol: LOC499607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760983.1 | XP_008759205.1 | 2 | 468 | ||

LOC689629
Genomic Map