siRNA Details
| Gene Symbol: LOC685667 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250713.1 | XP_006250775.1 | 1 | 135 | ||
| Gene Symbol: LOC100363333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250717.1 | XP_006250779.1 | 1 | 135 | ||

LOC685667
Genomic Map