siRNA Details
Gene Symbol: Dydc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252817.2 | XP_006252879.1 | 4 | 681 | ||
XM_006252818.2 | XP_006252880.1 | 4 | 341 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dydc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252817.2 | XP_006252879.1 | 4 | 681 | ||
XM_006252818.2 | XP_006252880.1 | 4 | 341 |