siRNA Details
| Gene Symbol: Smim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135252.1 | NP_001128724.1 | 3 | 115 | ||
| GenBank mRNA | BC158706.1 | - | 3 | 96 | 9030880 | |
| CB570183.1 | - | 120 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smim7| Gene Symbol: Smim7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135252.1 | NP_001128724.1 | 3 | 115 | ||
| GenBank mRNA | BC158706.1 | - | 3 | 96 | 9030880 | |
| CB570183.1 | - | 120 | ||||