siRNA Details
| Gene Symbol: LOC688459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109502.1 | NP_001102972.1 | 1 | 356 | ||
| GenBank mRNA | BC167028.1 | - | 1 | 340 | 9091474 | |
| FQ212996.1 | - | 1 | 355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC688459| Gene Symbol: LOC688459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109502.1 | NP_001102972.1 | 1 | 356 | ||
| GenBank mRNA | BC167028.1 | - | 1 | 340 | 9091474 | |
| FQ212996.1 | - | 1 | 355 | |||