siRNA Details
| Gene Symbol: Ccl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191092.1 | NP_001178021.1 | 3 | 223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl1| Gene Symbol: Ccl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191092.1 | NP_001178021.1 | 3 | 223 | ||