siRNA Details
| Gene Symbol: PLPPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014839.4 | NP_055654.2 | 6 | 1047 | ||
| XM_011542498.2 | XP_011540800.1 | 6 | 995 | |||
| GenBank mRNA | AB007924.3 | - | 6 | 1043 | ||
| AF357013.1 | - | 6 | 820 | |||
| AF541281.1 | - | 6 | 1054 | |||
| AK291369.1 | - | 6 | 847 | |||
| AL834390.1 | - | 5 | 560 | |||
| AY304518.1 | - | 6 | 995 | |||
| BC042963.1 | - | 6 | 865 | |||

PLPPR4
Genomic Map