siRNA Details
| Gene Symbol: Gpr84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109509.1 | NP_001102979.1 | 2 | 897 | ||
| XM_006242448.2 | XP_006242510.1 | 2 | 954 | |||
| XM_006242449.2 | XP_006242511.1 | 2 | 903 | |||
More Information
Set Membership: |
|

Gpr84
Genomic Map