siRNA Details
| Gene Symbol: Hdhd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109511.1 | NP_001102981.1 | 2 | 684 | ||
| GenBank mRNA | BC166898.1 | - | 2 | 681 | 6922018 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hdhd3| Gene Symbol: Hdhd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109511.1 | NP_001102981.1 | 2 | 684 | ||
| GenBank mRNA | BC166898.1 | - | 2 | 681 | 6922018 | |