siRNA Details
| Gene Symbol: Cd209f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001068161.5 | XP_001068161.4 | 8 | 1110 | ||
| XM_006248785.2 | XP_006248847.2 | 7 | 1026 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd209f| Gene Symbol: Cd209f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001068161.5 | XP_001068161.4 | 8 | 1110 | ||
| XM_006248785.2 | XP_006248847.2 | 7 | 1026 | |||