siRNA Details
| Gene Symbol: TECPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172631.2 | NP_001166102.1 | 7 | 1347 | ||
| NM_014844.4 | NP_055659.2 | 7 | 1347 | |||
| GenBank mRNA | AB002295.1 | - | 4 | 650 | ||
| AB002327.1 | - | 7 | 1296 | |||
| AK127721.1 | - | 7 | 1780 | |||
| BC136647.1 | - | 6 | 1144 | 9020637 | ||
| BC142667.1 | - | 7 | 1180 | 40148157 | ||
| BC142715.1 | - | 7 | 1180 | 40148843 | ||
| BC151230.1 | - | 7 | 1320 | 8860116 | ||

TECPR2
Genomic Map