siRNA Details
| Gene Symbol: Zbtb32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109517.1 | NP_001102987.1 | 1 | 507 | ||
| XM_006228793.2 | XP_006228855.1 | 2 | 1179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb32| Gene Symbol: Zbtb32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109517.1 | NP_001102987.1 | 1 | 507 | ||
| XM_006228793.2 | XP_006228855.1 | 2 | 1179 | |||