siRNA Details
| Gene Symbol: Tsga13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107851.1 | NP_001101321.1 | 5 | 561 | ||
| XM_006236230.2 | XP_006236292.1 | 3 | 295 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsga13| Gene Symbol: Tsga13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107851.1 | NP_001101321.1 | 5 | 561 | ||
| XM_006236230.2 | XP_006236292.1 | 3 | 295 | |||