siRNA Details
Gene Symbol: Ctps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134873.1 | NP_001128345.1 | 3 | 420 | ||
XM_006238781.2 | XP_006238843.1 | 3 | 413 | |||
GenBank mRNA | BC161954.1 | - | 3 | 398 | 9027103 | |
FQ210258.1 | - | 3 | 412 | |||
FQ217494.1 | - | 3 | 378 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
