siRNA Details
| Gene Symbol: LOC688869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145273.1 | NP_001138745.1 | 3 | 377 | ||
| XM_006228795.1 | XP_006228857.1 | 4 | 485 | |||
| GenBank mRNA | FQ217670.1 | - | 4 | 399 | ||
| FQ217813.1 | - | 4 | 395 | |||
| FQ218018.1 | - | 4 | 447 | |||
| FQ221178.1 | - | 4 | 443 | |||
| FQ222745.1 | - | 4 | 427 | |||
| FQ222968.1 | - | 4 | 445 | |||
| FQ223479.1 | - | 4 | 437 | |||
| FQ223722.1 | - | 4 | 405 | |||
| FQ223947.1 | - | 4 | 405 | |||
| FQ224063.1 | - | 4 | 450 | |||
| FQ224135.1 | - | 4 | 405 | |||
| FQ226525.1 | - | 4 | 438 | |||

LOC688869
Genomic Map