siRNA Details
Gene Symbol: Mrps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109518.1 | NP_001102988.1 | 2 | 282 | ||
GenBank mRNA | FQ228795.1 | - | 2 | 260 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mrps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109518.1 | NP_001102988.1 | 2 | 282 | ||
GenBank mRNA | FQ228795.1 | - | 2 | 260 |