siRNA Details
| Gene Symbol: Mrps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109518.1 | NP_001102988.1 | 2, 3 | 357 | ||
| GenBank mRNA | FQ210083.1 | - | 2, 3 | 312 | ||
| FQ228795.1 | - | 2, 3 | 335 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps16| Gene Symbol: Mrps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109518.1 | NP_001102988.1 | 2, 3 | 357 | ||
| GenBank mRNA | FQ210083.1 | - | 2, 3 | 312 | ||
| FQ228795.1 | - | 2, 3 | 335 | |||