siRNA Details
| Gene Symbol: Dcun1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134798.1 | NP_001128270.1 | 5, 6 | 627 | ||
| Gene Symbol: NEWGENE_1582994 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771413.1 | XP_008769635.1 | 5, 6 | 795 | ||

Dcun1d2
Genomic Map