siRNA Details
| Gene Symbol: Cd209c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248792.2 | XP_006248854.1 | 7 | 1717 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd209c| Gene Symbol: Cd209c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248792.2 | XP_006248854.1 | 7 | 1717 | ||