siRNA Details
| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001069014.4 | XP_001069014.1 | 3 | 270 | ||
| XM_006255409.2 | XP_006255471.1 | 3 | 245 | |||
| XM_006255410.2 | XP_006255472.1 | 3 | 246 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gypa| Gene Symbol: Gypa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001069014.4 | XP_001069014.1 | 3 | 270 | ||
| XM_006255409.2 | XP_006255471.1 | 3 | 245 | |||
| XM_006255410.2 | XP_006255472.1 | 3 | 246 | |||