siRNA Details
| Gene Symbol: Ccdc84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001060013.5 | XP_001060013.1 | 7 | 674 | ||
| XM_008766177.1 | XP_008764399.1 | 6 | 651 | |||
| XM_008766178.1 | XP_008764400.1 | 7 | 695 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc84| Gene Symbol: Ccdc84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_001060013.5 | XP_001060013.1 | 7 | 674 | ||
| XM_008766177.1 | XP_008764399.1 | 6 | 651 | |||
| XM_008766178.1 | XP_008764400.1 | 7 | 695 | |||